siRNA Details
| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006960.1 | NP_001006961.1 | 3 | 421 | ||
| GenBank mRNA | BC083713.1 | - | 3 | 421 | 7190675 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtfp1| Gene Symbol: Mtfp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006960.1 | NP_001006961.1 | 3 | 421 | ||
| GenBank mRNA | BC083713.1 | - | 3 | 421 | 7190675 | |