siRNA Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006961.1 | NP_001006962.1 | 3 | 342 | ||
| XM_006251199.1 | XP_006251261.1 | 2 | 1905 | |||
| XM_006251200.2 | XP_006251262.1 | 3 | 1258 | |||
| GenBank mRNA | AB167521.1 | - | 3 | 342 | ||

Osm
Genomic Map