siRNA Details
| Gene Symbol: Rhbdd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013875.1 | NP_001013897.1 | 6 | 1222 | ||
| XM_006251204.2 | XP_006251266.1 | 7 | 1319 | |||
| XM_006251205.2 | XP_006251267.1 | 6 | 1109 | |||
| XM_006251206.2 | XP_006251268.1 | 7 | 1152 | |||
| XM_006251207.2 | XP_006251269.1 | 6 | 1110 | |||
| XM_006251208.2 | XP_006251270.1 | 7 | 1259 | |||
| XM_006251209.2 | XP_006251271.1 | 6 | 1302 | |||
| XM_006251210.2 | XP_006251272.1 | 5 | 1098 | |||
| GenBank mRNA | BC081912.1 | - | 6 | 1222 | 7112018 | |

Rhbdd3
Genomic Map