siRNA Details
| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013050.1 | NP_001013068.1 | 3 | 311 | ||
| GenBank mRNA | BC079267.1 | - | 3 | 311 | 7133662 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pold2| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013050.1 | NP_001013068.1 | 3 | 311 | ||
| GenBank mRNA | BC079267.1 | - | 3 | 311 | 7133662 | |