siRNA Details
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013050.1 | NP_001013068.1 | 4 | 455 | ||
GenBank mRNA | BC079267.1 | - | 4 | 455 | 7133662 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pold2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013050.1 | NP_001013068.1 | 4 | 455 | ||
GenBank mRNA | BC079267.1 | - | 4 | 455 | 7133662 |