siRNA Details
| Gene Symbol: RGD1309870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106018.1 | NP_001099488.1 | 5, 6 | 537 | ||
| XM_006251478.2 | XP_006251540.1 | 8, 9 | 1611 | |||
| XM_006251479.2 | XP_006251541.1 | 7, 8 | 1569 | |||
| XM_006251481.2 | XP_006251543.1 | 7, 8 | 812 | |||
| XM_006251482.2 | XP_006251544.1 | 7, 8 | 604 | |||

RGD1309870
Genomic Map