siRNA Details
| Gene Symbol: Ddx56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004211.1 | NP_001004211.1 | 10, 11 | 1480 | ||
| XM_006251437.2 | XP_006251499.1 | 11, 12 | 1415 | |||
| GenBank mRNA | BC078919.1 | - | 10, 11 | 1480 | 7096512 | |
| FQ212430.1 | - | 1401 | ||||

Ddx56
Genomic Map