siRNA Details
| Gene Symbol: PHLDA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003311.3 | NP_003302.1 | 2 | 687 | ||
| GenBank mRNA | AF001294.1 | - | 2 | 687 | ||
| AF019953.1 | - | 2 | 648 | |||
| AF035444.1 | - | 2 | 687 | |||
| AK223027.1 | - | 2 | 687 | |||
| BC005034.1 | - | 2 | 677 | 3954346 | ||

PHLDA2
Genomic Map