siRNA Details
Gene Symbol: LOC103693776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770394.1 | XP_008768616.1 | 18 | 7541 | ||
XM_008770395.1 | XP_008768617.1 | 17 | 3467 | |||
XM_008770396.1 | XP_008768618.1 | 19 | 6817 | |||
XM_008770397.1 | XP_008768619.1 | 17 | 7463 | |||
XM_008770398.1 | XP_008768620.1 | 18 | 7445 | |||
XM_008770399.1 | XP_008768621.1 | 18 | 6739 | |||
XM_008770400.1 | XP_008768622.1 | 19 | 6721 | |||
XM_008770401.1 | XP_008768623.1 | 16 | 3470 | |||
XM_008770402.1 | XP_008768624.1 | 16 | 3599 | |||
XM_008770403.1 | XP_008768625.1 | 17 | 3403 | |||
XM_008770404.1 | XP_008768626.1 | 18 | 6643 |
Gene Symbol: Zmiz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100507.1 | NP_001093977.1 | 3305 | |||
XM_006251439.2 | XP_006251501.1 | 6739 | ||||
XM_006251440.2 | XP_006251502.1 | 6721 | ||||
XM_008770262.1 | XP_008768484.1 | 7541 | ||||
XM_008770263.1 | XP_008768485.1 | 3467 | ||||
XM_008770264.1 | XP_008768486.1 | 7463 | ||||
XM_008770265.1 | XP_008768487.1 | 7445 | ||||
XM_008770266.1 | XP_008768488.1 | 3470 | ||||
XM_008770267.1 | XP_008768489.1 | 3599 | ||||
XM_008770268.1 | XP_008768490.1 | 3403 | ||||
XM_008770269.1 | XP_008768491.1 | 6643 | ||||
GenBank mRNA | BC079401.1 | - | 1155 | |||
BC099192.1 | - | 3 | 325 |
