siRNA Details
| Gene Symbol: Pno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199083.2 | NP_954514.2 | 2 | 352 | ||
| GenBank mRNA | AY344057.1 | - | 2 | 352 | ||
| BC088091.1 | - | 2 | 329 | 7325613 | ||
| FQ215167.1 | - | 2 | 319 | |||
| FQ232331.1 | - | 2 | 332 | |||

Pno1
Genomic Map