siRNA Details
| Gene Symbol: Cep68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_214106.7 | XP_214106.4 | 2 | 356 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep68| Gene Symbol: Cep68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_214106.7 | XP_214106.4 | 2 | 356 | ||