siRNA Details
| Gene Symbol: Ugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024743.1 | NP_001019914.1 | 4, 5 | 698 | ||
| XM_006251538.1 | XP_006251600.1 | 4, 5 | 650 | |||
| GenBank mRNA | BC097369.1 | - | 4, 5 | 698 | 7443949 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugp2| Gene Symbol: Ugp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024743.1 | NP_001019914.1 | 4, 5 | 698 | ||
| XM_006251538.1 | XP_006251600.1 | 4, 5 | 650 | |||
| GenBank mRNA | BC097369.1 | - | 4, 5 | 698 | 7443949 | |