siRNA Details
| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013876.2 | NP_001013898.2 | 3 | 1237 | ||
| XM_006251574.2 | XP_006251636.2 | 3 | 1237 | |||
| XM_008770425.1 | XP_008768647.1 | 3 | 1237 | |||
| GenBank mRNA | BC079233.1 | - | 3 | 1131 | 7132584 | |

Fam161a
Genomic Map