siRNA Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106024.1 | NP_001099494.1 | 20, 21 | 2374 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde6b| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106024.1 | NP_001099494.1 | 20, 21 | 2374 | ||