siRNA Details
| Gene Symbol: Tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007619.1 | NP_001007620.1 | 3 | 521 | ||
| GenBank mRNA | AY539871.1 | - | 433 | |||
| BC079452.1 | - | 3 | 521 | 7121068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmed5| Gene Symbol: Tmed5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007619.1 | NP_001007620.1 | 3 | 521 | ||
| GenBank mRNA | AY539871.1 | - | 433 | |||
| BC079452.1 | - | 3 | 521 | 7121068 | ||