siRNA Details
| Gene Symbol: TTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282500.1 | NP_001269429.1 | 6 | 755 | ||
| NM_003314.2 | NP_003305.1 | 6 | 758 | |||
| GenBank mRNA | AK222641.1 | - | 6 | 709 | ||
| AK222984.1 | - | 6 | 703 | |||
| AK315262.1 | - | 6 | 713 | |||
| BC000942.3 | - | 5 | 663 | 3446109 | ||
| U46570.1 | - | 6 | 685 | |||

TTC1
Genomic Map