siRNA Details
| Gene Symbol: RGD1306063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106026.2 | NP_001099496.1 | 3 | 139 | ||
| XM_006251730.2 | XP_006251792.1 | 3 | 173 | |||
| XM_006251731.2 | XP_006251793.1 | 3 | 170 | |||
| XM_006251732.2 | XP_006251794.1 | 3 | 191 | |||
| XM_006251733.2 | XP_006251795.1 | 3 | 175 | |||
| XM_006251736.2 | XP_006251798.1 | 3 | 173 | |||
| GenBank mRNA | BC168755.1 | - | 3 | 139 | 6887151 | |

RGD1306063
Genomic Map