siRNA Details
| Gene Symbol: Cdkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251791.2 | XP_006251853.1 | 7, 8 | 646 | ||
| XM_006251792.2 | XP_006251854.1 | 7, 8 | 643 | |||
| GenBank mRNA | BC167026.1 | - | 6, 7 | 560 | 8375927 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn3| Gene Symbol: Cdkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251791.2 | XP_006251853.1 | 7, 8 | 646 | ||
| XM_006251792.2 | XP_006251854.1 | 7, 8 | 643 | |||
| GenBank mRNA | BC167026.1 | - | 6, 7 | 560 | 8375927 | |