siRNA Details
| Gene Symbol: Cdkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106028.1 | NP_001099498.1 | 2 | 141 | ||
| XM_006251791.2 | XP_006251853.1 | 2 | 172 | |||
| XM_006251792.2 | XP_006251854.1 | 2 | 172 | |||
| GenBank mRNA | BC167026.1 | - | 1 | 86 | 8375927 | |

Cdkn3
Genomic Map