siRNA Details
| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135802.1 | NP_001129274.1 | 17 | 2304 | ||
| XM_006251794.2 | XP_006251856.1 | 17 | 2377 | |||
| XM_008770506.1 | XP_008768728.1 | 17 | 2370 | |||
| GenBank mRNA | BC162031.1 | - | 17 | 2304 | 9101206 | |

Dlgap5
Genomic Map