siRNA Details
| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106030.1 | NP_001099500.1 | 8 | 688 | ||
| XM_006251856.2 | XP_006251918.1 | 6 | 532 | |||
| GenBank mRNA | BC161972.1 | - | 5 | 2258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parp2| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106030.1 | NP_001099500.1 | 8 | 688 | ||
| XM_006251856.2 | XP_006251918.1 | 6 | 532 | |||
| GenBank mRNA | BC161972.1 | - | 5 | 2258 | ||