siRNA Details
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100510.1 | NP_001093980.1 | 2 | 301 | ||
GenBank mRNA | BC087140.1 | - | 2 | 225 | ||
BC158592.1 | - | 2 | 301 | 8362956 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100510.1 | NP_001093980.1 | 2 | 301 | ||
GenBank mRNA | BC087140.1 | - | 2 | 225 | ||
BC158592.1 | - | 2 | 301 | 8362956 |