siRNA Details
| Gene Symbol: Osgep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100510.1 | NP_001093980.1 | 4 | 569 | ||
| GenBank mRNA | BC087140.1 | - | 4 | 493 | ||
| BC158592.1 | - | 4 | 569 | 8362956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osgep| Gene Symbol: Osgep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100510.1 | NP_001093980.1 | 4 | 569 | ||
| GenBank mRNA | BC087140.1 | - | 4 | 493 | ||
| BC158592.1 | - | 4 | 569 | 8362956 | ||