siRNA Details
| Gene Symbol: Olr1640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000099.1 | NP_001000099.1 | 1 | 274 | ||
| Gene Symbol: Olr1639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000833.1 | NP_001000833.1 | 1 | 274 | ||

Olr1640
Genomic Map