siRNA Details
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106034.2 | NP_001099504.2 | 4 | 733 | ||
GenBank mRNA | CX569344.1 | - | 3 | 249 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cmtm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106034.2 | NP_001099504.2 | 4 | 733 | ||
GenBank mRNA | CX569344.1 | - | 3 | 249 |