siRNA Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106036.2 | NP_001099506.1 | 1 | 61 | ||
| XM_006251935.2 | XP_006251997.1 | 3 | 340 | |||
| XM_006251937.2 | XP_006251999.1 | 3 | 320 | |||
| XM_008770672.1 | XP_008768894.1 | 2 | 243 | |||

Nrl
Genomic Map