siRNA Details
| Gene Symbol: Tssk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271325.1 | NP_001258254.1 | 4 | 1198 | ||
| XM_006251938.2 | XP_006252000.1 | 4 | 7506 | |||
| XM_006251939.2 | XP_006252001.1 | 4 | 7476 | |||
| XM_008770674.1 | XP_008768896.1 | 5 | 7440 | |||
| GenBank mRNA | BC079202.1 | - | 4 | 1254 | 7131733 | |

Tssk4
Genomic Map