siRNA Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007621.1 | NP_001007622.1 | 9 | 1270 | ||
| XM_006251946.2 | XP_006252008.1 | 9 | 1135 | |||
| XM_006251947.2 | XP_006252009.1 | 10 | 1203 | |||
| XM_006251948.2 | XP_006252010.1 | 10 | 1181 | |||
| GenBank mRNA | BC079263.1 | - | 9 | 1270 | 7133513 | |
| FQ211258.1 | - | 7 | 981 | |||

Dhrs1
Genomic Map