siRNA Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007621.1 | NP_001007622.1 | 5 | 850 | ||
| XM_006251946.2 | XP_006252008.1 | 5 | 715 | |||
| XM_006251947.2 | XP_006252009.1 | 6 | 783 | |||
| XM_006251948.2 | XP_006252010.1 | 6 | 761 | |||
| GenBank mRNA | BC079263.1 | - | 5 | 850 | 7133513 | |

Dhrs1
Genomic Map