siRNA Details
| Gene Symbol: Nop9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106040.1 | NP_001099510.1 | 7 | 1430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nop9| Gene Symbol: Nop9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106040.1 | NP_001099510.1 | 7 | 1430 | ||