siRNA Details
| Gene Symbol: Ctsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106041.1 | NP_001099511.1 | 2 | 290 | ||
| XM_006252041.1 | XP_006252103.1 | 2 | 202 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsg| Gene Symbol: Ctsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106041.1 | NP_001099511.1 | 2 | 290 | ||
| XM_006252041.1 | XP_006252103.1 | 2 | 202 | |||