siRNA Details
| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175757.2 | NP_786933.1 | 7 | 815 | ||
| XM_006252058.2 | XP_006252120.1 | 5 | 1336 | |||
| GenBank mRNA | AY040223.1 | - | 7 | 815 | ||
| BC078685.1 | - | 7 | 789 | 7110482 | ||

Cryl1
Genomic Map