siRNA Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106042.1 | NP_001099512.1 | 17 | 2750 | ||
| XM_006252062.2 | XP_006252124.1 | 17 | 2754 | |||
| XM_006252063.2 | XP_006252125.1 | 17 | 2626 | |||
| GenBank mRNA | FQ225683.1 | - | 5 | 823 | ||
| FQ231595.1 | - | 5 | 823 | |||
| FQ234166.1 | - | 5 | 823 | |||

Xpo4
Genomic Map