siRNA Details
| Gene Symbol: Cab39l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011917.1 | NP_001011917.1 | 6, 7 | 811 | ||
| XM_006252098.2 | XP_006252160.1 | 7, 8 | 915 | |||
| XM_006252099.2 | XP_006252161.1 | 8, 9 | 999 | |||
| XM_006252100.2 | XP_006252162.1 | 9, 10 | 1195 | |||
| XM_006252101.2 | XP_006252163.1 | 8, 9 | 1111 | |||
| XM_008770742.1 | XP_008768964.1 | 8, 9 | 1008 | |||
| XM_008770743.1 | XP_008768965.1 | 5, 6 | 1405 | |||
| GenBank mRNA | BC083684.1 | - | 6, 7 | 811 | 7188949 | |

Cab39l
Genomic Map