siRNA Details
| Gene Symbol: Tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106044.1 | NP_001099514.1 | 5, 6 | 519 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdh| Gene Symbol: Tdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106044.1 | NP_001099514.1 | 5, 6 | 519 | ||