siRNA Details
| Gene Symbol: Sox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106045.1 | NP_001099515.1 | 1 | 304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sox7| Gene Symbol: Sox7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106045.1 | NP_001099515.1 | 1 | 304 | ||