siRNA Details
| Gene Symbol: Adamdec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106046.1 | NP_001099516.1 | 10 | 1070 | ||
| XM_006252229.2 | XP_006252291.1 | 11 | 1128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamdec1| Gene Symbol: Adamdec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106046.1 | NP_001099516.1 | 10 | 1070 | ||
| XM_006252229.2 | XP_006252291.1 | 11 | 1128 | |||