siRNA Details
| Gene Symbol: LOC102550885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252233.2 | XP_006252295.1 | 3 | 291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102550885| Gene Symbol: LOC102550885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252233.2 | XP_006252295.1 | 3 | 291 | ||