siRNA Details
| Gene Symbol: Adam28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181693.1 | NP_859044.1 | 4, 5 | 341 | ||
| XM_008770820.1 | XP_008769042.1 | 4, 5 | 352 | |||
| GenBank mRNA | AY283187.1 | - | 4, 5 | 341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam28| Gene Symbol: Adam28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181693.1 | NP_859044.1 | 4, 5 | 341 | ||
| XM_008770820.1 | XP_008769042.1 | 4, 5 | 352 | |||
| GenBank mRNA | AY283187.1 | - | 4, 5 | 341 | ||