siRNA Details
| Gene Symbol: Npm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203340.1 | NP_976085.1 | 5 | 496 | ||
| XM_006252261.2 | XP_006252323.1 | 6 | 1306 | |||
| XM_008770824.1 | XP_008769046.1 | 2 | 1170 | |||
| XM_008770825.1 | XP_008769047.1 | 3 | 1223 | |||

Npm2
Genomic Map