siRNA Details
| Gene Symbol: Itm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006963.1 | NP_001006964.1 | 5 | 772 | ||
| GenBank mRNA | BC083735.1 | - | 5 | 772 | 7191589 | |
| FQ210227.1 | - | 5 | 802 | |||
| FQ212539.1 | - | 2 | 149 | |||
| FQ212925.1 | - | 5 | 729 | |||
| FQ218718.1 | - | 5 | 771 | |||
| FQ219378.1 | - | 2 | 135 | |||
| FQ229426.1 | - | 5 | 771 | |||
| FQ230065.1 | - | 5 | 744 | |||

Itm2b
Genomic Map