siRNA Details
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770921.1 | XP_008769143.1 | 4822 | |||
| GenBank mRNA | BC083871.1 | - | 4 | 496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa8| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770921.1 | XP_008769143.1 | 4822 | |||
| GenBank mRNA | BC083871.1 | - | 4 | 496 | ||