siRNA Details
| Gene Symbol: Slitrk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106057.1 | NP_001099527.1 | 2 | 2806 | ||
| XM_006252443.2 | XP_006252505.1 | 2 | 3039 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slitrk6| Gene Symbol: Slitrk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106057.1 | NP_001099527.1 | 2 | 2806 | ||
| XM_006252443.2 | XP_006252505.1 | 2 | 3039 | |||