siRNA Details
| Gene Symbol: Thoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106059.1 | NP_001099529.1 | 5 | 903 | ||
| GenBank mRNA | BC168852.1 | - | 5 | 886 | 7443262 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thoc3| Gene Symbol: Thoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106059.1 | NP_001099529.1 | 5 | 903 | ||
| GenBank mRNA | BC168852.1 | - | 5 | 886 | 7443262 | |