siRNA Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011918.1 | NP_001011918.1 | 9, 10 | 1097 | ||
| XM_008770980.1 | XP_008769202.1 | 9, 10 | 1343 | |||
| GenBank mRNA | BC083812.1 | - | 9, 10 | 1097 | 7110191 | |

Anxa11
Genomic Map