siRNA Details
| Gene Symbol: DNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005336.2 | NP_001005336.1 | 8 | 1126 | ||
| NM_001288737.1 | NP_001275666.1 | 8 | 1126 | |||
| NM_001288738.1 | NP_001275667.1 | 8 | 1126 | |||
| NM_001288739.1 | NP_001275668.1 | 8 | 1126 | |||
| NM_004408.3 | NP_004399.2 | 8 | 1126 | |||
| XM_005251763.2 | XP_005251820.1 | 8 | 1126 | |||
| XM_005251768.2 | XP_005251825.1 | 8 | 1126 | |||
| XM_005251769.2 | XP_005251826.1 | 8 | 1126 | |||
| XM_006716992.2 | XP_006717055.1 | 8 | 1126 | |||
| XM_006716993.1 | XP_006717056.1 | 8 | 1126 | |||
| XM_011518335.2 | XP_011516637.1 | 8 | 1126 | |||
| XM_011518336.2 | XP_011516638.1 | 8 | 1126 | |||
| XM_017014369.1 | XP_016869858.1 | 8 | 1126 | |||
| XM_017014370.1 | XP_016869859.1 | 8 | 1126 | |||
| XM_017014371.1 | XP_016869860.1 | 8 | 1126 | |||
| XM_017014372.1 | XP_016869861.1 | 8 | 1126 | |||
| XM_017014373.1 | XP_016869862.1 | 8 | 1126 | |||
| GenBank mRNA | AB209124.1 | - | 7 | 811 | ||
| AK316235.1 | - | 8 | 1102 | |||
| BC050279.1 | - | 8 | 1102 | 5284480 | ||

DNM1
Genomic Map