siRNA Details
Gene Symbol: Slmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106060.1 | NP_001099530.1 | 8 | 1808 | ||
XM_006252541.2 | XP_006252603.1 | 7 | 770 | |||
XM_006252542.2 | XP_006252604.1 | 7 | 770 | |||
XM_006252543.2 | XP_006252605.1 | 7 | 770 | |||
XM_006252544.2 | XP_006252606.1 | 7 | 770 | |||
XM_006252545.2 | XP_006252607.1 | 7 | 770 | |||
XM_006252546.2 | XP_006252608.1 | 7 | 770 | |||
XM_006252548.2 | XP_006252610.1 | 7 | 770 | |||
XM_006252549.2 | XP_006252611.1 | 7 | 770 | |||
XM_006252550.2 | XP_006252612.1 | 7 | 770 | |||
XM_006252551.2 | XP_006252613.1 | 7 | 770 | |||
XM_006252553.2 | XP_006252615.1 | 8 | 1826 | |||
XM_008770981.1 | XP_008769203.1 | 4 | 349 | |||
GenBank mRNA | BC168941.1 | - | 8 | 1757 | 7104670 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_032008, AK163630, AK166396, AK164911, AK141597, and others.
Panther Classification:
