siRNA Details
| Gene Symbol: Slmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106060.1 | NP_001099530.1 | 16 | 2651 | ||
| XM_006252541.2 | XP_006252603.1 | 18 | 1778 | |||
| XM_006252542.2 | XP_006252604.1 | 18 | 1778 | |||
| XM_006252543.2 | XP_006252605.1 | 17 | 1727 | |||
| XM_006252544.2 | XP_006252606.1 | 17 | 1727 | |||
| XM_006252545.2 | XP_006252607.1 | 17 | 1715 | |||
| XM_006252546.2 | XP_006252608.1 | 17 | 1715 | |||
| XM_006252548.2 | XP_006252610.1 | 16 | 1664 | |||
| XM_006252549.2 | XP_006252611.1 | 16 | 1664 | |||
| XM_006252550.2 | XP_006252612.1 | 17 | 1655 | |||
| XM_006252551.2 | XP_006252613.1 | 15 | 1613 | |||
| XM_006252553.2 | XP_006252615.1 | 15 | 2546 | |||
| XM_008770981.1 | XP_008769203.1 | 15 | 1357 | |||
| GenBank mRNA | BC168941.1 | - | 15 | 2477 | 7104670 | |
| FQ217977.1 | - | 4 | 239 | |||

Slmap
Genomic Map