siRNA Details
| Gene Symbol: Selk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207589.3 | NP_997472.2 | 2 | 152 | ||
| GenBank mRNA | AY319924.1 | - | 2 | 75 | ||
| BC088086.1 | - | 2 | 96 | 7326199 | ||
| CF979418.1 | - | 2 | 156 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Selk| Gene Symbol: Selk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207589.3 | NP_997472.2 | 2 | 152 | ||
| GenBank mRNA | AY319924.1 | - | 2 | 75 | ||
| BC088086.1 | - | 2 | 96 | 7326199 | ||
| CF979418.1 | - | 2 | 156 | |||