siRNA Details
| Gene Symbol: Rft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135866.1 | NP_001129338.1 | 1, 2 | 121 | ||
| XM_006252581.2 | XP_006252643.1 | 1, 2 | 132 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rft1| Gene Symbol: Rft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135866.1 | NP_001129338.1 | 1, 2 | 121 | ||
| XM_006252581.2 | XP_006252643.1 | 1, 2 | 132 | |||